Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4,4'-Bis(decyloxy)-3-methylazobenzene 95.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| Quantity | 500 mg |
| Formula Weight | 508.79 |
| Percent Purity | ≥95.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,4′-Bis(decyloxy)-3-methylazobenzene |
Safety and Handling
| TSCA | No |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?