Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(diethylphosphonomethyl)biphenyl 98.0+%, TCI America™
SDP

Catalog No. B19235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19235G 5 g
B19231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19235G Supplier TCI America Supplier No. B19235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17919-34-5
Molecular Formula C22H32O6P2
Molecular Weight (g/mol) 454.44
MDL Number MFCD01321144
InChI Key DRCOGQJUVZRNSQ-UHFFFAOYSA-N
PubChem CID 635132
IUPAC Name 1-(diethoxyphosphorylmethyl)-4-[4-(diethoxyphosphorylmethyl)phenyl]benzene
SMILES CCOP(=O)(CC1=CC=C(C=C1)C2=CC=C(C=C2)CP(=O)(OCC)OCC)OCC

Specifications

Color White
Quantity 5 g
Synonym [4,4′-Biphenylylenebis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl [4,4′-Biphenylylenebis(methylene)]bisphosphonate
Formula Weight 454.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(diethylphosphonomethyl)biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.