Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(hydroxymethyl)-2,2'-bipyridine 95.0+%, TCI America™
SDP

Catalog No. B35111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35111G 1 g
B35115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35111G Supplier TCI America Supplier No. B35111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109073-77-0
Molecular Formula C12H12N2O2
Molecular Weight (g/mol) 216.24
MDL Number MFCD06202808
InChI Key RRXGRDMHWYLJSY-UHFFFAOYSA-N
PubChem CID 11160239
IUPAC Name [2-[4-(hydroxymethyl)pyridin-2-yl]pyridin-4-yl]methanol
SMILES C1=CN=C(C=C1CO)C2=NC=CC(=C2)CO

Specifications

Melting Point 174°C
Color White
Quantity 1 g
Synonym 2,2′C-Bipyridine-4,4′C-dimethanol, 4,4′C-Bis(hydroxymethyl)-2,2′C-bipyridyl
Formula Weight 216.24
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(hydroxymethyl)-2,2′-bipyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.