Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester | 470478-90-1 | 98.5% | C21H33BN2O4 | 100 G
SDP

Catalog No. 5000434931
Encompass_Preferred

4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester is a PROTAC linker utilized in the synthesis of CST905. This compound functions as part of Proteolysis Targeting Chimeras (PROTACs), which are molecules designed to selectively degrade target proteins by harnessing the intracellular ubiquitin-proteasome system. PROTACs achieve this by using two ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other targets a specific protein. This product is for research use only.

  • Functions as a PROTAC linker
  • Used in the synthesis of CST905
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
  • Purity is 98.50%
  • Molecular weight is 388.31
  • Appearance is solid, white to light yellow

Catalog No. 50-004-34931 Supplier Medchemexpress LLC Supplier No. HY78378100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester is a PROTAC linker utilized in the synthesis of CST905. This compound functions as part of Proteolysis Targeting Chimeras (PROTACs), which are molecules designed to selectively degrade target proteins by harnessing the intracellular ubiquitin-proteasome system. PROTACs achieve this by using two ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other targets a specific protein. This product is for research use only.

  • Functions as a PROTAC linker
  • Used in the synthesis of CST905
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
  • Purity is 98.50%
  • Molecular weight is 388.31
  • Appearance is solid, white to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.