Learn More
Medchemexpress LLC 4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester | 470478-90-1 | 98.5% | C21H33BN2O4 | 5 G
SDP
Catalog No. 5000433323
Encompass_Preferred
4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester is a PROTAC linker that can be used in the synthesis of CST905 (HY-175435). PROTACs (Proteolysis-targeting chimeras) are molecules that contain two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. For research use only. MedChemExpress does not sell to patients.
  • PROTAC linker
  • Used in the synthesis of CST905
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-004-33323 Supplier Medchemexpress LLC Supplier No. HY783785G
4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester is a PROTAC linker that can be used in the synthesis of CST905 (HY-175435). PROTACs (Proteolysis-targeting chimeras) are molecules that contain two different ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. For research use only. MedChemExpress does not sell to patients.
  • PROTAC linker
  • Used in the synthesis of CST905
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only