Learn More
Medchemexpress LLC 4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester | 470478-90-1 | 98.5% | C21H33BN2O4 | 50 G
SDP
Catalog No. 5000432615
Encompass_Preferred
4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester is a PROTAC linker that can be utilized in the synthesis of CST905. PROTACs consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Functions as a PROTAC linker
  • Can be used in the synthesis of certain PROTAC compounds
  • Designed to target E3 ubiquitin ligases and specific proteins of interest
  • Utilizes the ubiquitin-proteasome system for targeted protein degradation
Catalog No. 50-004-32615 Supplier Medchemexpress LLC Supplier No. HY7837850G
4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester is a PROTAC linker that can be utilized in the synthesis of CST905. PROTACs consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting the protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Functions as a PROTAC linker
  • Can be used in the synthesis of certain PROTAC compounds
  • Designed to target E3 ubiquitin ligases and specific proteins of interest
  • Utilizes the ubiquitin-proteasome system for targeted protein degradation