Learn More
Medchemexpress LLC 4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester | 470478-90-1 | 98.5% | C21H33BN2O4 | 500 MG
SDP
Catalog No. 5000434863
Encompass_Preferred
This product, 4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester, functions as a PROTAC linker, utilized in the synthesis of compounds such as CST905. PROTACs are designed to leverage the intracellular ubiquitin-proteasome system, enabling the selective degradation of target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein. This compound appears as a white to light yellow solid and is designated for research purposes only.
  • Functions as a PROTAC linker
  • Used in the synthesis of CST905
  • Exploits ubiquitin-proteasome system for selective protein degradation
  • Appears as a white to light yellow solid
  • For research use only
Catalog No. 50-004-34863 Supplier Medchemexpress LLC Supplier No. HY78378500MG
This product, 4-(4-Boc-piperazinemethyl)phenylboronic acid pinacol ester, functions as a PROTAC linker, utilized in the synthesis of compounds such as CST905. PROTACs are designed to leverage the intracellular ubiquitin-proteasome system, enabling the selective degradation of target proteins by linking a ligand for an E3 ubiquitin ligase with a ligand for the target protein. This compound appears as a white to light yellow solid and is designated for research purposes only.
  • Functions as a PROTAC linker
  • Used in the synthesis of CST905
  • Exploits ubiquitin-proteasome system for selective protein degradation
  • Appears as a white to light yellow solid
  • For research use only