Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4-(4-Bromophenoxy)phenyl)methanol

Catalog No. JRD06781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD06781G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD06781G Supplier Sigma Aldrich Supplier No. JRD06781G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Diethyl allyl phosphate can be prepared by reacting ethanol and pocl3.it can undergo free radical polymerization to form a flame retardant polymer, poly deapdeap may be used as a flame retardant additive in the preparation of rubber wood/polymer composite
  • It can also be used for the stereoselective synthesis of cis- or trans-3-vinyl-α-lactam compounds and also for the fabrication of amphoteric surfaces on silicone substrates

Specifications

Molecular Formula C13H11BrO2
Linear Formula C13H11BrO2
Quantity 5 g
MDL Number MFCD11190398
Molecular Weight (g/mol) 279.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.