Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4'-Bromophenyl)-4-hydroxypiperidine 98.0+%, TCI America™
SDP

Catalog No. B171910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B171910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B171910G Supplier TCI America Supplier No. B171910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57988-58-6
Molecular Formula C11H15BrNO
Molecular Weight (g/mol) 257.15
MDL Number MFCD00067034
InChI Key QNLXJYQUWCNYBH-UHFFFAOYSA-O
Synonym 4-4-bromophenyl piperidin-4-ol, 4-4'-bromophenyl-4-hydroxypiperidine, 4-4-bromophenyl-4-piperidinol, 4-4-bromophenyl-4-hydroxy piperidine, 4-piperidinol, 4-4-bromophenyl, 4-piperidinol,4-4-bromophenyl, 4-4-bromophenyl-4-hydroxypiperidine, 4-4-bromophenyl-piperidin-4-ol, 4-4-bromo-phenyl-piperidin-4-ol, 4-4-bromophenyl-4-piperidinol #
PubChem CID 93911
IUPAC Name 4-(4-bromophenyl)-4-hydroxypiperidin-1-ium
SMILES OC1(CC[NH2+]CC1)C1=CC=C(Br)C=C1

Specifications

Melting Point 167°C
Quantity 10 g
Formula Weight 256.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4′-Bromophenyl)-4-hydroxypiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.