Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Bromophenyl)morpholine 98.0+%, TCI America™
SDP

Catalog No. B48905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48905G 5 g
B489025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48905G Supplier TCI America Supplier No. B48905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30483-75-1
Molecular Formula C10H12BrNO
Molecular Weight (g/mol) 242.116
MDL Number MFCD04112483
InChI Key UJTKZWNRUPTHSB-UHFFFAOYSA-N
PubChem CID 11622851
IUPAC Name 4-(4-bromophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)Br

Specifications

Melting Point 115°C
Quantity 5 g
Formula Weight 242.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.