Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde 95.0+%, TCI America™
SDP

Catalog No. B4922200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4922200MG 200 mg
B49221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4922200MG Supplier TCI America Supplier No. B4922200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 733744-98-4
Molecular Formula C20H16BrNO
Molecular Weight (g/mol) 366.258
MDL Number MFCD16658914
InChI Key SDKPEZBGWABWKD-UHFFFAOYSA-N
Synonym 4-[(4-Bromophenyl)(4-methylphenyl)amino]benzaldehyde
PubChem CID 58544534
IUPAC Name 4-(N-(4-bromophenyl)-4-methylanilino)benzaldehyde
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C=O)C3=CC=C(C=C3)Br

Specifications

Melting Point 117°C
Quantity 200 mg
Formula Weight 366.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(4-Bromophenyl)(p-tolyl)amino]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.