Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Bromophenyl)phenol, 97%, Thermo Scientific™

Catalog No. AC338360250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC338360250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC338360250 Supplier Thermo Scientific Chemicals Supplier No. 338360250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29558-77-8
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
InChI Key ARUBXNBYMCVENE-UHFFFAOYSA-N
Synonym 4-bromo-4'-hydroxybiphenyl, 4'-bromo-1,1'-biphenyl-4-ol, 4-4-bromophenyl phenol, 4'-bromobiphenyl-4-ol, 4-hydroxy-4'-bromobiphenyl, 1,1'-biphenyl-4-ol, 4'-bromo, 4-bromo-4-hydroxybiphenyl, pubchem23025
PubChem CID 95093
IUPAC Name 4-(4-bromophenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)Br)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 163°C to 168°C
Color Cream or White
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (Br)C6H4(C6H4OH)
Quantity 25 g
Formula Weight 249.11
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 4-(4-Bromophenyl)phenol, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.