Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Bromophenyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B46631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B46631G Supplier TCI America Supplier No. B46631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent suitable for materials science applications.

Chemical Identifiers

CAS 39795-60-3
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD00956748
InChI Key GYJBDJGUNDKZKO-UHFFFAOYSA-N
PubChem CID 616283
IUPAC Name 4-(4-bromophenyl)pyridine
SMILES C1=CC(=CC=C1C2=CC=NC=C2)Br

Specifications

Melting Point 131°C
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.