Learn More
Medchemexpress LLC 4-(4-carbamoylphenoxy)benzamide | 6336-34-1 | 99.8% | 256.26 g/mol | C14H12N2O3 | 5MG
SDP
Catalog No. 5000236961
Encompass_Preferred
OUL35 (NSC39047) is a small-molecule inhibitor selective for ARTD10 (PARP10) with a reported IC50 of 329 nM. It is supplied as a high-purity solid; solutions in DMSO and premade solution formats are available. Common uses include biochemical and cellular assays to probe PARP10 activity and mono-ADP-ribosylation signaling.
  • Selective ARTD10 (PARP10) inhibitor; IC50 329 nM.
  • High purity suitable for analytical and biological studies.
  • Available as solid and as DMSO solution formats.
  • Molecular weight 256.26 g/mol; formula C14H12N2O3.
  • Store protected from light; follow solvent storage instructions for solutions.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-36961 Supplier Medchemexpress LLC Supplier No. HY1235125MG
OUL35 (NSC39047) is a small-molecule inhibitor selective for ARTD10 (PARP10) with a reported IC50 of 329 nM. It is supplied as a high-purity solid; solutions in DMSO and premade solution formats are available. Common uses include biochemical and cellular assays to probe PARP10 activity and mono-ADP-ribosylation signaling.
  • Selective ARTD10 (PARP10) inhibitor; IC50 329 nM.
  • High purity suitable for analytical and biological studies.
  • Available as solid and as DMSO solution formats.
  • Molecular weight 256.26 g/mol; formula C14H12N2O3.
  • Store protected from light; follow solvent storage instructions for solutions.
  • Intended for research use only; not for human or clinical use.