Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorophenoxy)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. C20065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20065G Supplier TCI America Supplier No. C20065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61343-99-5
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.66
MDL Number MFCD01631895
InChI Key BLCXBCYVCDPFEU-UHFFFAOYSA-N
Synonym 4-4-chlorophenoxy benzaldehyde, 4-chloro-4'-formyldiphenyl ether, acmc-1b4h0, 4-4-chlorophoxy benzaldehyde, blcxbcyvcdpfeu-uhfffaoysa, 4-4-chloranylphenoxy benzaldehyde, benzaldehyde,4-4-chlorophenoxy
PubChem CID 3851764
IUPAC Name 4-(4-chlorophenoxy)benzaldehyde
SMILES ClC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1

Specifications

Melting Point 55°C
Color Yellow
Boiling Point 181°C
Quantity 5 g
Formula Weight 232.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenoxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.