Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%

Catalog No. ac109930010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109930010 1 g Glass bottle
AC109930050 5 g Glass bottle
AC109930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC109930010 Supplier Thermo Scientific Chemicals Supplier No. 109930010
Only null left
Add to Cart

CAS: 51304-61-1 | C11H12ClN·HCl | 230.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51304-61-1
MDL Number MFCD00012753
InChI Key NGDJUNABWNTULX-UHFFFAOYSA-N
Synonym 4-4-chlorophenyl-1,2,3,6-tetrahydropyridine hydrochloride, 4-4-chlorophenyl-1,2,3,6-tetrahydropyridinium chloride, 4-4-chlorophenyl-1,2,5,6-tetrahydropyridine, chloride, acmc-1alu0, 4-4-chlorophenyl-1,2,3,6-tetrahydropyridine hcl, 4-4-chloro-phenyl-1,2,3,6-tetrahydro-pyridine hydrochloride, 4-4-chlorophenyl-1?2?3?6-tetrahydropyridine hydrochloride, 4-4-chlorophenyl-1,2,3,6-tetrahydropyridine monohydrochloride, pyridine, 4-4-chlorophenyl-1,2,3,6-tetrahydro-, hydrochloride
PubChem CID 2723612
IUPAC Name 4-(4-chlorophenyl)-1,2,3,6-tetrahydropyridine;hydrochloride
SMILES C1CNCC=C1C2=CC=C(C=C2)Cl.Cl

Specifications

Melting Point 199.0°C to 204.0°C
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97%
Molecular Formula C11H12ClN·HCl
Molecular Weight (g/mol) 230.14
Formula Weight 230.14
Percent Purity 97%
Chemical Name or Material 4-(4-Chlorophenyl)-1,2,3,6-tetrahydropyridine hydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Toxic in contact with skin.
May cause respiratory irritation.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 257-126-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.