Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(4-Chlorophenyl)-4-hydroxypiperidine

Catalog No. C660565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C660565G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C660565G Supplier Sigma Aldrich Supplier No. C660565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-(4-Chlorophenyl)-4-hydroxypiperidine

Specifications

CAS 39512-49-7
Melting Point 137°C to 140°C (lit.)
Molecular Formula C11H14ClNO
Linear Formula C11H14ClNO
MDL Number MFCD00006001
Quantity 250 mg
Synonym Haloperidol metabolite I; 4-(4-Chlorophenyl)-4-hydroxypiperidine; 4-(4-Chlorophenyl)-4-piperidinol
Molecular Weight (g/mol) 211.69
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.