Learn More
Medchemexpress LLC 4-(4-Chlorophenyl)-6-(4-hydroxy-3-methoxyphenyl)pyrimidin-2(1H)-one | 2172879-52-4 | >99.9% | 328.06 | 50 MG
SDP
Catalog No. 5000437772
Encompass_Preferred
LIT-927 is a small molecule that acts as a neutraligand by binding to and neutralizing the chemokine CXCL12. Developed as an alternative strategy to inhibitors for CXCL12's cognate receptors CXCR4 or CXCR7, it functions as a potential anti-inflammatory agent.
  • Locally and orally active
  • Functions as a CXCL12 neutraligand
  • Exhibits anti-inflammatory effects
  • Binds to the specific receptor CXCR4 with a Ki of 267 nM for CXCL12 binding
Catalog No. 50-004-37772 Supplier Medchemexpress LLC Supplier No. HY11270950MG
LIT-927 is a small molecule that acts as a neutraligand by binding to and neutralizing the chemokine CXCL12. Developed as an alternative strategy to inhibitors for CXCL12's cognate receptors CXCR4 or CXCR7, it functions as a potential anti-inflammatory agent.
  • Locally and orally active
  • Functions as a CXCL12 neutraligand
  • Exhibits anti-inflammatory effects
  • Binds to the specific receptor CXCR4 with a Ki of 267 nM for CXCL12 binding