Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorophenyl)morpholine 98.0+%, TCI America™
SDP

Catalog No. C20825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20825G Supplier TCI America Supplier No. C20825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70291-67-7
Molecular Formula C10H12ClNO
Molecular Weight (g/mol) 197.662
InChI Key KDWPYZPZUPWJGB-UHFFFAOYSA-N
PubChem CID 10420251
IUPAC Name 4-(4-chlorophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)Cl

Specifications

Melting Point 73°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 197.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.