Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(4-chlorophenyl)-piperidine hydrochloride

Catalog No. CDS015248 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS015248 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS015248 Supplier Sigma Aldrich Supplier No. CDS015248100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-methyl-1-pyrroline, a monocyclic imine, is a pyrroline derivative
  • It is a five-membered heterocyclic compound having various biological and pharmacological applications
  • It is formed during the rh(i) complexes (containing n,n-donor ligands and n,p-donor ligand) immobilized on glassy carbon electrode surfaces catalyzed intramolecular hydroamination of 4-pentyn-1-amine
  • It reacts with with 2-oxopropanal to afford acetyl-1-pyrroline (ap).2-methyl-1-pyrroline (2-mpn) may be used in the enantioselective enzymatic synthesis of (r)- and (s)-2-methylpyrroline in the presence of whole-cells catalysts isolated from strptomyces sp
  • Strains gf3587 and gf3546 respectively.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 381055.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

Molecular Formula C11H15Cl2N
Linear Formula C11H15Cl2N
MDL Number MFCD02178903
Quantity 25 mL
Molecular Weight (g/mol) 232.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.