Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorophenyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. C19355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19355G Supplier TCI America Supplier No. C19355G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82222-74-0
Molecular Formula C10H12ClNO2S
Molecular Weight (g/mol) 245.72
MDL Number MFCD07366283
InChI Key IQIXFXLKPNFQGQ-UHFFFAOYSA-N
Synonym 4-(1,1-Dioxothiomorpholino)phenyl Chloride
PubChem CID 12932466
IUPAC Name 4-(4-chlorophenyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES ClC1=CC=C(C=C1)N1CCS(=O)(=O)CC1

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 5 g
Formula Weight 245.72
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Chlorophenyl)thiomorpholine 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.