Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-cis-Stilbenedicarboxylic acid, 95%

Catalog No. AAL0436403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0436403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0436403 Supplier Thermo Scientific Chemicals Supplier No. L0436403
  • $80.60
    $68.65 / Each of 1

    Save $11.95

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 133005-88-6 | C16H12O4 | 268.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133005-88-6
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00013994
InChI Key SBBQDUFLZGOASY-UPHRSURJSA-N
Synonym 4-z-2-4-carboxyphenyl ethenyl benzoic acid, cis-stilbene-4,4'-dicarboxylic acid, z-4,4'-ethene-1,2-diyl dibenzoic acid, cis-4,4'-dicarboxystilbene, z-4,4'-stilbene dicarboxylic acid, z-stilbene-4,4'-dicarboxylic acid, 4,4'-cis-stilbenedicarboxylic acid
PubChem CID 18647961
IUPAC Name 4-[(Z)-2-(4-carboxyphenyl)ethenyl]benzoic acid
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)C(=O)O)C(=O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 1978660
Formula Weight 268.27
Percent Purity 95%
Chemical Name or Material 4,4'-cis-Stilbenedicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.