Learn More
Medchemexpress LLC 4-[[4-cyclohexyl-N-[(3-methylsulfonylphenyl)carbamoyl]anilino]methyl]-N-(2H-tetrazol-5-yl)benzamide | 307986-98-7 | 98.4% | 573.67 g/mol | C29H31N7O4S | 100MG
SDP
Supplier:  Medchemexpress LLC HY124622100MG
Encompass_Preferred
NNC-0640 is a small-molecule negative allosteric modulator (NAM) of the human glucagon receptor (GCGR) characterized for preclinical research. It shows an IC50 of 69.2 nM against human GCGR and is supplied at high purity for biochemical and cellular studies. Intended for research use only; not for human or veterinary use.
  • Negative allosteric modulator of human glucagon receptor (GCGR).
  • IC50 = 69.2 nM in human GCGR assays.
  • High purity (98.4%) suitable for biochemical and cellular studies.
  • Molecular weight 573.67 g/mol.
  • Provided as a 100 mg package for laboratory use.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-36147