Learn More
Medchemexpress LLC 4-[[(4-cyclohexylphenyl)[[[3-(methylsulfonyl)phenyl]amino]carbonyl]amino]methyl]-N-2H-tetrazol-5-yl- | 307986-98-7 | >98.0% | 573.67 g/mol | C29H31N7O4S | 25MG
SDP
Supplier:  Medchemexpress LLC HY12462225MG
Encompass_Preferred
NNC-0640 (CAS 307986-98-7) is a research small-molecule negative allosteric modulator of the human glucagon receptor (GCGR) used to study glucagon signaling and metabolic regulation. It is supplied as a solid and is reported with high purity, making it suitable for in vitro pharmacology and biochemical assays.
Catalog No. 50-002-37319