Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-DDT Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USPP1801
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-1801 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-PP-1801 Supplier Agilent Technologies Supplier No. PP1801
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

CAS 50-29-3, 67-56-1
CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,4,4'-DDT
Molecular Formula C14H9Cl5
InChI Key YVGGHNCTFXOJCH-UHFFFAOYSA-N
Element Methanol,4,4'-DDT
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)Cl)C(Cl)(Cl)Cl)Cl
Solvent or Matrix Methanol
IUPAC Name 1-chloro-4-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
Molecular Weight (g/mol) 354.476
Quantity 1 x 1 mL
PubChem CID 3036
ChEBI CHEBI:16130
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 4, 4'-DDT Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.