Learn More
Medchemexpress LLC 4',4'''-Di-O-methylisochamaejasmin | 1620921-68-7 | 98.4% | 570.54 | 5 MG
SDP
Catalog No. 5000425269
Encompass_Preferred
4',4'''-Di-O-methylisochamaejasmin, a flavonoid and biflavone originating from Thymelaeaceae Stellera chamaejasme L., exhibits anti-cancer effects. This product is intended for research use only.
  • Exhibits anti-cancer effects
  • Intended for research use only
  • Classified as a flavonoid and biflavone
  • Originates from plants, specifically Thymelaeaceae Stellera chamaejasme L.
  • Recommended storage for powder: -20°C for 3 years, or 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month
Catalog No. 50-004-25269 Supplier Medchemexpress LLC Supplier No. HYN92255MG
4',4'''-Di-O-methylisochamaejasmin, a flavonoid and biflavone originating from Thymelaeaceae Stellera chamaejasme L., exhibits anti-cancer effects. This product is intended for research use only.
  • Exhibits anti-cancer effects
  • Intended for research use only
  • Classified as a flavonoid and biflavone
  • Originates from plants, specifically Thymelaeaceae Stellera chamaejasme L.
  • Recommended storage for powder: -20°C for 3 years, or 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month