Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4,4'-Di-tert-butyl-2,2'-bipyridine)bis[(2-pyridinyl)phenyl]iridium(III) Hexafluorophosphate 85.0+%, TCI America™
SDP

Catalog No. D4887200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4887200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4887200MG Supplier TCI America Supplier No. D4887200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 676525-77-2
Molecular Formula C40H40F6IrN4P
Molecular Weight (g/mol) 913.969
InChI Key VCIVELSSYHAWGC-UHFFFAOYSA-N
PubChem CID 44239994
IUPAC Name 4-tert-butyl-2-(4-tert-butylpyridin-2-yl)pyridine;iridium(3+);2-phenylpyridine;hexafluorophosphate
SMILES CC(C)(C)C1=CC(=NC=C1)C2=NC=CC(=C2)C(C)(C)C.C1=CC=C([C-]=C1)C2=CC=CC=N2.C1=CC=C([C-]=C1)C2=CC=CC=N2.F[P-](F)(F)(F)(F)F.[Ir+3]

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 913.97
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (4,4′-Di-tert-butyl-2,2′-bipyridine)bis[(2-pyridinyl)phenyl]iridium(III) Hexafluorophosp
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.