Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Di-tert-butylbenzophenone 95.0+%, TCI America™
SDP

Catalog No. D53081G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D53081G Supplier TCI America Supplier No. D53081G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15796-82-4
Molecular Formula C21H26O
Molecular Weight (g/mol) 294.438
MDL Number MFCD01319937
InChI Key YNPFOBWIQVHZMO-UHFFFAOYSA-N
Synonym Bis[4-(tert-butyl)phenyl]methanone
PubChem CID 2758048
IUPAC Name bis(4-tert-butylphenyl)methanone
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C(C)(C)C

Specifications

Melting Point 134°C
Color White
Boiling Point 158°C
Quantity 1 g
Formula Weight 294.44
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Di-tert-butylbenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.