Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Diaminodiphenylmethane, MilliporeSigma™ Supelco™

Catalog No. 111019131 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9131 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9131 Supplier MilliporeSigma Supelco Supplier No. 31640250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4,4′-Diaminodiphenylmethane is a curing agent for epoxy resins. It is prepared from formaldehyde aniline when heated with aniline hydrochloride and aniline.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-Methylenedianiline
CAS 101-77-9
Flash Point 230°C
Physical Form Neat
Linear Formula C13H14N2
Technique HPLC
MDL Number MFCD00007914
Melting Point 88°C to 92°C
Synonym 4,4 ′-Methylenedianiline; MDA
Type Analytical Standard
Industry Type Food and Beverage
Molecular Weight (g/mol) 198.26
Quantity 250 mg
Formula Weight 198.26
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.