Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diamyloxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. D10905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10905G Supplier TCI America Supplier No. D10905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21470-41-7
Molecular Formula C22H30O2
Molecular Weight (g/mol) 326.48
MDL Number MFCD00059432
InChI Key XTKHESQIKTWMCD-UHFFFAOYSA-N
PubChem CID 629825
IUPAC Name 4,4'-bis(pentyloxy)-1,1'-biphenyl
SMILES CCCCCOC1=CC=C(C=C1)C1=CC=C(OCCCCC)C=C1

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 326.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diamyloxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.