Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromo-2,2'-bipyridine, 98%

Catalog No. AC454900050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454900050 5 g
AC454900010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454900050 Supplier Thermo Scientific Chemicals Supplier No. 454900050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18511-71-2 | C10H6Br2N2 | 313.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18511-71-2
Molecular Formula C10H6Br2N2
Molecular Weight (g/mol) 313.98
InChI Key KIIHBDSNVJRWFY-UHFFFAOYSA-N
Synonym 4,4'-dibromo-2,2'-bipyridine, 4-bromo-2-4-bromopyridin-2-yl pyridine, 4,4'-dibromo-2,2'-bipyridyl, 2,2'-bipyridine, 4,4'-dibromo, bpbb01, acmc-1bo83, 4,4'-dibromo-2,2-bipyridine, 4,4'-bromo-2,2'-bipyridine, 4,4'-dibromo-2,2'-dipyridine, 4,4'-dibromine-2,2'-bipyridine
PubChem CID 15569987
IUPAC Name 4-bromo-2-(4-bromopyridin-2-yl)pyridine
SMILES C1=CN=C(C=C1Br)C2=NC=CC(=C2)Br

Specifications

Melting Point 140.0°C to 143.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 313.98
Percent Purity 98%
Chemical Name or Material 4,4'-Dibromo-2,2'-bipyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.