Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibutoxyazoxybenzene, TCI America™
SDP

Catalog No. D10961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10961G Supplier TCI America Supplier No. D10961G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17051-01-3
Molecular Formula C20H26N2O3
Molecular Weight (g/mol) 342.439
MDL Number MFCD00048767
InChI Key DWRXFZDFCLDVQG-UHFFFAOYSA-N
PubChem CID 86924
IUPAC Name (4-butoxyphenyl)-(4-butoxyphenyl)imino-oxidoazanium
SMILES CCCCOC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OCCCC)[O-]

Specifications

Quantity 1 g
Formula Weight 342.44
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibutoxyazoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.