Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibutoxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. D108225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D108225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D108225G Supplier TCI America Supplier No. D108225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39800-63-0
Molecular Formula C20H26O2
Molecular Weight (g/mol) 298.426
MDL Number MFCD00059419
InChI Key IBCVAKLNEYUUBM-UHFFFAOYSA-N
PubChem CID 604312
IUPAC Name 1-butoxy-4-(4-butoxyphenyl)benzene
SMILES CCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)OCCCC

Specifications

Color White
Quantity 25 g
Formula Weight 298.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibutoxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.