Learn More
Cayman Chemical 4,4'-Dicyanobenzhydrol, Purity: ≥98%, CAS: 134521-16-7, 10 mg
SDP
Catalog No. 502980898
Encompass_Preferred
An analytical reference standard that is a metabolite of letrozole; has been used as a marker of letrozole use in athletes; intended for research and forensic applications
Catalog No. 50-298-0898 Supplier Cayman Chemical Supplier No. 2131610 MG
An analytical reference standard that is a metabolite of letrozole; has been used as a marker of letrozole use in athletes; intended for research and forensic applications