Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,4-difluoro-8-(4-iodophenyl)-1,3,5,7-tetramethyl-4-bora-3a,4a-diaza-s-indacene | 250734-47-5 | MFCD29905036 | 98.5% | 450.1 g/mol | C19H18BF2IN2 | 100 MG
SDP

Catalog No. 5000281349
Encompass_Preferred

8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is a fluorescent BODIPY dye used as a probe for GFP-labeled muscarinic M1 receptors. It offers strong fluorescence and photostability, making it suitable for biological imaging and analytical applications.

  • High fluorescence intensity suitable for imaging applications.
  • Photostable dye for reliable signal over time.
  • Molecular formula C19H18BF2IN2.
  • Molecular weight 450.1 g/mol.
  • Purity approximately 98.5% as reported.
  • Reported use as a probe for GFP-labeled muscarinic M1 receptor.
  • Available in 100 MG quantities.

Catalog No. 50-002-81349 Supplier Medchemexpress LLC Supplier No. HYW248119100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is a fluorescent BODIPY dye used as a probe for GFP-labeled muscarinic M1 receptors. It offers strong fluorescence and photostability, making it suitable for biological imaging and analytical applications.

  • High fluorescence intensity suitable for imaging applications.
  • Photostable dye for reliable signal over time.
  • Molecular formula C19H18BF2IN2.
  • Molecular weight 450.1 g/mol.
  • Purity approximately 98.5% as reported.
  • Reported use as a probe for GFP-labeled muscarinic M1 receptor.
  • Available in 100 MG quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.