Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorodiphenylmethane, 99%, Thermo Scientific™

Catalog No. AC243280100
Change view
Click to view available options
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC243280100 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243280100 Supplier Thermo Scientific Chemicals Supplier No. 243280100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 457-68-1
CAS Min % 98.5
CAS Max % 100.0
Density 1.1500g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C13H10F2
Refractive Index 1.5345 to 1.5365 (20°C, 589nm)
Quantity 10mL
MDL Number MFCD00000360
Beilstein 05, IV, 1846
Synonym bis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethane, 1,1'-methylenebis 4-fluorobenzene, 1-fluoro-4-4-fluorophenyl methyl benzene, benzene, 1,1'-methylenebis 4-fluoro, 4-fluoro-1-4-fluorophenyl methyl benzene, 4,4'-difluorodiphenylamine, bis-4-fluorophenyl methane, 4,4'-difluorodiphenyl methane, 1,1-methylenebis 4-fluorobenzene
InChI Key DXQVFHQUHOFROC-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)F)F
IUPAC Name 1-fluoro-4-[(4-fluorophenyl)methyl]benzene
Molecular Weight (g/mol) 204.22
PubChem CID 96094
Formula Weight 204.22
Percent Purity ≥98.5%
Chemical Name or Material 4, 4'-Difluorodiphenylmethane, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-274-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.