Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihexyloxybiphenyl 98.0+%, TCI America™
SDP

Catalog No. D10895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D10895G Supplier TCI America Supplier No. D10895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142450-58-6
Molecular Formula C24H34O2
Molecular Weight (g/mol) 354.534
MDL Number MFCD00059438
InChI Key NPALUNJGWGOZQN-UHFFFAOYSA-N
PubChem CID 604320
IUPAC Name 1-hexoxy-4-(4-hexoxyphenyl)benzene
SMILES CCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)OCCCCCC

Specifications

Melting Point 128°C
Color White
Quantity 5 g
Formula Weight 354.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihexyloxybiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.