Learn More
Medchemexpress LLC 4,4'-dihydroxybiphenyl | 92-88-6 | MFCD00002348 | 100.0% | 186.21 g/mol | C12H10O2 | 100 G
SDP
Catalog No. 5000302691
Encompass_Preferred
4,4'-Dihydroxybiphenyl is a phenolic biphenyl compound provided for laboratory and life-science research. It functions as a reagent and synthetic intermediate used in the preparation of plastics and polyether liquid crystals, and as an antioxidant additive in rubber and latex formulations.
  • High purity (99.95% reported).
  • Molecular formula C12H10O2 and molecular weight 186.21 g/mol.
  • Solid form, white to light-yellow powder suitable for bench-scale synthesis.
  • Versatile reagent and intermediate for organic synthesis.
  • Applicable as an antioxidant in rubber, latex, and related materials.
Catalog No. 50-003-02691 Supplier Medchemexpress LLC Supplier No. HYW016189100G
Add to Cart
Add to Cart
4,4'-Dihydroxybiphenyl is a phenolic biphenyl compound provided for laboratory and life-science research. It functions as a reagent and synthetic intermediate used in the preparation of plastics and polyether liquid crystals, and as an antioxidant additive in rubber and latex formulations.
  • High purity (99.95% reported).
  • Molecular formula C12H10O2 and molecular weight 186.21 g/mol.
  • Solid form, white to light-yellow powder suitable for bench-scale synthesis.
  • Versatile reagent and intermediate for organic synthesis.
  • Applicable as an antioxidant in rubber, latex, and related materials.