Learn More
4,4'-Dihydroxybiphenyl, 99%
Description
4,4'-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,4′-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 279°C to 285°C |
| Odor | Odorless |
| Quantity | 100 g |
| Beilstein | 1908886 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 186.21 |
| Percent Purity | 99% |
| Chemical Name or Material | 4,4'-Dihydroxybiphenyl |
Safety and Handling
| Hazard Category | H312-H315-H319-H335 |
| Hazard Statement | GHS H Statement H312-H315-H319-H335 Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 202-200-5 |
| RTECSNumber | DV4725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.