Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihydroxydiphenyl Ether, MP Biomedicals

Catalog No. ICN205693 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205693 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205693 Supplier MP Biomedicals, Inc Supplier No. 0520569301

Chemical Identifiers

CAS 1965-09-9
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.21
InChI Key NZGQHKSLKRFZFL-UHFFFAOYSA-N
Synonym 4,4'-oxydiphenol, 4,4'-dihydroxydiphenyl ether, phenol, 4,4'-oxybis, 4-4-hydroxyphenoxy phenol, p,p'-oxydiphenol, phenol, 4,4'-oxydi, p,p'-oxybisphenol, 4,4'-oxybisphenol, bis 4-hydroxyphenyl ether, unii-7l83kgh26s
PubChem CID 16069
IUPAC Name 4-(4-hydroxyphenoxy)phenol
SMILES C1=CC(=CC=C1O)OC2=CC=C(C=C2)O

Specifications

Quantity 1 g
Chemical Name or Material 4, 4«-Dihydroxydiphenyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.