Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxybenzhydrol, MP Biomedicals

Catalog No. ICN227187 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227187 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227187 Supplier MP Biomedicals, Inc Supplier No. 0522718710

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 728-87-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008410
InChI Key ZODAOVNETBTTJX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh
PubChem CID 69768
IUPAC Name bis(4-methoxyphenyl)methanol
SMILES COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1

Specifications

Quantity 10 g
Formula Weight 244.29
Chemical Name or Material 4, 4«-Dimethoxybenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.