Learn More
Medchemexpress LLC 4,4'-Dimethyl-2,2'-bipyridine | 1134-35-6 | 184.24 | 10 G
SDP
Catalog No. 5000446424
Encompass_Preferred
4,4'-Dimethyl-2,2'-bipyridine is a biochemical reagent suitable for life science related research. It is used for the determination of ferrous and cyanide compounds.
  • Starting material for the synthesis of blue-emitting molecules.
  • Can be used as a biological material or organic compound.
  • Excited at 43% quantum field and 450 nm.
Catalog No. 50-004-46424 Supplier Medchemexpress LLC Supplier No. HYW00200310G
4,4'-Dimethyl-2,2'-bipyridine is a biochemical reagent suitable for life science related research. It is used for the determination of ferrous and cyanide compounds.
  • Starting material for the synthesis of blue-emitting molecules.
  • Can be used as a biological material or organic compound.
  • Excited at 43% quantum field and 450 nm.