Learn More
AdipoGen 4 4-Dimethyl-2 2-dipyridyl
SDP
Catalog No. 502561643
Encompass_Preferred
Chemical. CAS 1134-35-6. Formula C12H12N2. MW 184.2. Synthetic. Building block and intermediate for synthesis. Used in the synthesis of a series of o-phenanthroline-substituted rutheniumII complexes and luminescent molecules.
Catalog No. 50-256-1643 Supplier AdipoGen Supplier No. CDXB0213G025
Chemical. CAS 1134-35-6. Formula C12H12N2. MW 184.2. Synthetic. Building block and intermediate for synthesis. Used in the synthesis of a series of o-phenanthroline-substituted rutheniumII complexes and luminescent molecules.