Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane 95.0+%, TCI America™
SDP

Catalog No. D52001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52001G 1 g
D52005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52001G Supplier TCI America Supplier No. D52001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57280-22-5
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD16621162
InChI Key GEKNCWQQNMEIMS-UHFFFAOYNA-N
PubChem CID 11094747
IUPAC Name 4,4-dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
SMILES CC1(C)OCC2OC2CO1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 1 g
Formula Weight 144.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.