Learn More
Medchemexpress LLC 4-[4-[(dimethylamino)methyl]-2,5-dimethoxyphenyl]-2-methyl-1,2-dihydro-2,7-naphthyridin-1-one | 1883429-22-8 | MFCD30182324 | 98.7% | 353.41 Da | C20H23N3O3 | 50 MG
SDP
Catalog No. 5000212677
Encompass_Preferred
BI-9564 is a small-molecule chemical probe that selectively inhibits BRD9 and, to a lesser extent, BRD7 bromodomains. It is cell-permeable, exhibits high affinity for BRD9, and has been used in both in vitro and in vivo studies to probe BAF complex biology and bromodomain function.
Catalog No. 50-002-12677 Supplier Medchemexpress LLC Supplier No. HY10035250MG
BI-9564 is a small-molecule chemical probe that selectively inhibits BRD9 and, to a lesser extent, BRD7 bromodomains. It is cell-permeable, exhibits high affinity for BRD9, and has been used in both in vitro and in vivo studies to probe BAF complex biology and bromodomain function.