Learn More
Medchemexpress LLC 4-(4-((dimethylamino)methyl)-2,5-dimethoxyphenyl)-2-methyl-2,7-naphthyridin-1(2H)-one | 1883429-22-8 | 98.7% | 353.41 | C20H23N3O3 | 10 MG
SDP
Catalog No. 5000213276
Encompass_Preferred
BI-9564 is a potent, selective, and cell-permeable inhibitor of BRD9 and BRD7 bromodomains used as a chemical probe in epigenetics research. It exhibits low-nanomolar affinity for BRD9, displays selectivity versus BET family bromodomains, and is supplied as a white to off-white solid for laboratory use.
  • Potent inhibition of BRD9 with low-nanomolar affinity.
  • Selectivity for BRD9/BRD7 over BET family bromodomains.
  • Cell-permeable for use in cellular assays.
  • High purity suitable for research applications.
  • Supplied as a solid with defined storage recommendations.
Catalog No. 50-002-13276 Supplier Medchemexpress LLC Supplier No. HY10035210MG
BI-9564 is a potent, selective, and cell-permeable inhibitor of BRD9 and BRD7 bromodomains used as a chemical probe in epigenetics research. It exhibits low-nanomolar affinity for BRD9, displays selectivity versus BET family bromodomains, and is supplied as a white to off-white solid for laboratory use.
  • Potent inhibition of BRD9 with low-nanomolar affinity.
  • Selectivity for BRD9/BRD7 over BET family bromodomains.
  • Cell-permeable for use in cellular assays.
  • High purity suitable for research applications.
  • Supplied as a solid with defined storage recommendations.