Learn More
Medchemexpress LLC 4-[4-[(dimethylamino)methyl]-3,5-dimethoxyphenyl]-2-methyl-2,7-naphthyridin-1(2H)-one | 1883429-21-7 | 100.0% | 353.41 Da | C20H23N3O3 | 50 MG
SDP
Catalog No. 5000214352
Encompass_Preferred
BI-7273 is a selective, cell-permeable bromodomain inhibitor used as a research tool to probe BRD9 and BRD7 biology. It demonstrates potent biochemical activity and is provided as a powdered reagent for laboratory assays.
  • Selective, cell-permeable inhibitor of BRD9 and BRD7.
  • Potent biochemical activity: IC50 19 nM (BRD9) and 117 nM (BRD7).
  • Characterized compound: CAS 1883429-21-7; formula C20H23N3O3.
  • Supplied as a powder suitable for in vitro and in vivo research.
  • High purity appropriate for biochemical and cellular studies.
Catalog No. 50-002-14352 Supplier Medchemexpress LLC Supplier No. HY10035150MG
BI-7273 is a selective, cell-permeable bromodomain inhibitor used as a research tool to probe BRD9 and BRD7 biology. It demonstrates potent biochemical activity and is provided as a powdered reagent for laboratory assays.
  • Selective, cell-permeable inhibitor of BRD9 and BRD7.
  • Potent biochemical activity: IC50 19 nM (BRD9) and 117 nM (BRD7).
  • Characterized compound: CAS 1883429-21-7; formula C20H23N3O3.
  • Supplied as a powder suitable for in vitro and in vivo research.
  • High purity appropriate for biochemical and cellular studies.