Learn More
Medchemexpress LLC 4-[4-[(dimethylamino)methyl]-3,5-dimethoxyphenyl]-2-methyl-2,7-naphthyridin-1(2H)-one | 1883429-21-7 | >98.0% | 353.4 Da | C20H23N3O3 | 10 MG
SDP
Catalog No. 5000214347
Encompass_Preferred
BI-7273 is a selective, cell-permeable small-molecule inhibitor of the BRD9 bromodomain that also inhibits BRD7 with lower potency. It binds BRD9 with a reported Kd of ~15 nM and an AlphaScreen IC50 of 19 nM, while BRD7 AlphaScreen IC50 is reported as 117 nM. Reported CAS number is 1883429-21-7 and molecular weight is 353.4 Da. It is used as a research tool in epigenetic and leukemia studies.
Catalog No. 50-002-14347 Supplier Medchemexpress LLC Supplier No. HY10035110MG
BI-7273 is a selective, cell-permeable small-molecule inhibitor of the BRD9 bromodomain that also inhibits BRD7 with lower potency. It binds BRD9 with a reported Kd of ~15 nM and an AlphaScreen IC50 of 19 nM, while BRD7 AlphaScreen IC50 is reported as 117 nM. Reported CAS number is 1883429-21-7 and molecular weight is 353.4 Da. It is used as a research tool in epigenetic and leukemia studies.