Learn More
Medchemexpress LLC 4-[4-[(dimethylamino)methyl]-3,5-dimethoxyphenyl]-2-methyl-2,7-naphthyridin-1(2H)-one | 1883429-21-7 | ≥98.0% | 353.41 | C20H23N3O3 | 25 MG
SDP
Catalog No. 5000214572
Encompass_Preferred
BI-7273 is a potent, cell-permeable small-molecule inhibitor of bromodomain-containing proteins BRD9 and BRD7, developed for research use in epigenetics and oncology. It demonstrates low-nanomolar biochemical potency and high binding affinity, and is suitable as a tool compound for biochemical, cellular, and in vivo studies.
  • Potent biochemical activity (BRD9 IC50 ~19 nM; BRD7 IC50 ~117 nM).
  • High binding affinity (BRD9 Kd ~0.75 nM; BRD7 Kd ~0.3 nM).
  • Cell-permeable and active in cellular assays.
  • Suitable for in vitro and in vivo experiments.
  • Soluble in DMSO at 10 mg/mL; ultrasonic may be required.
  • Stable as a powder when stored at -20°C; follow supplier storage recommendations.
Catalog No. 50-002-14572 Supplier Medchemexpress LLC Supplier No. HY10035125MG
BI-7273 is a potent, cell-permeable small-molecule inhibitor of bromodomain-containing proteins BRD9 and BRD7, developed for research use in epigenetics and oncology. It demonstrates low-nanomolar biochemical potency and high binding affinity, and is suitable as a tool compound for biochemical, cellular, and in vivo studies.
  • Potent biochemical activity (BRD9 IC50 ~19 nM; BRD7 IC50 ~117 nM).
  • High binding affinity (BRD9 Kd ~0.75 nM; BRD7 Kd ~0.3 nM).
  • Cell-permeable and active in cellular assays.
  • Suitable for in vitro and in vivo experiments.
  • Soluble in DMSO at 10 mg/mL; ultrasonic may be required.
  • Stable as a powder when stored at -20°C; follow supplier storage recommendations.