Learn More
Medchemexpress LLC 4-[4-[(dimethylamino)methyl]-3,5-dimethoxyphenyl]-2-methyl-2,7-naphthyridin-1(2H)-one | 1883429-21-7 | MFCD30489736 | 99.8% | 353.41 g·mol⁻¹ | C20H23N3O3 | 100 MG
SDP
Catalog No. 5000214334
Encompass_Preferred
BI-7273 is a potent, selective, and cell-permeable small-molecule inhibitor of the BRD9 bromodomain intended for biochemical and cellular research. It displays low-nanomolar biochemical potency against BRD9, measurable activity versus BRD7, and is provided as a high-purity solid for laboratory use.
  • Selective inhibitor of BRD9 bromodomain with low-nanomolar potency.
  • Measurable activity against BRD7, supporting comparative studies.
  • Demonstrated low-nanomolar Kd and IC50 values in biochemical assays.
  • Cell-permeable and active in multiple cellular models.
  • Supplied as a high-purity solid with defined storage recommendations.
  • Suitable for biochemical, cellular, and structural biology applications.
Catalog No. 50-002-14334 Supplier Medchemexpress LLC Supplier No. HY100351100MG
Add to Cart
Add to Cart
BI-7273 is a potent, selective, and cell-permeable small-molecule inhibitor of the BRD9 bromodomain intended for biochemical and cellular research. It displays low-nanomolar biochemical potency against BRD9, measurable activity versus BRD7, and is provided as a high-purity solid for laboratory use.
  • Selective inhibitor of BRD9 bromodomain with low-nanomolar potency.
  • Measurable activity against BRD7, supporting comparative studies.
  • Demonstrated low-nanomolar Kd and IC50 values in biochemical assays.
  • Cell-permeable and active in multiple cellular models.
  • Supplied as a high-purity solid with defined storage recommendations.
  • Suitable for biochemical, cellular, and structural biology applications.